Tisk…

4-Amino-2-methylpyridine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine

OR6421-5G OR6421-25G OR6421-100G
APOSOR6421-5GEA 705 CZK
APOSOR6421-5G APOSOR6421-25G APOSOR6421-100G
4-Amino-2-methylpyridine 97%
4-Amino-2-methylpyridine
Vzorec: C₆H₈N₂
MW: 108,14 g/mol
Bod tání: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00186509
CAS číslo: 18437-58-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR