Tisk…

(R)-(-)-2-Hydroxyisovaleric acid 97%

Dodavatel: Apollo Scientific

Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid

OR6367-1G OR6367-5G
APOSOR6367-1GEA 7280 CZK
APOSOR6367-1G APOSOR6367-5G
(R)-(-)-2-Hydroxyisovaleric acid 97%
(R)-(-)-2-Hydroxyisovaleric acid
Vzorec: C₅H₁₀O₃
MW: 118,13 g/mol
Bod varu: 124…125 °C/13mmHg
Bod tání: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00066442
CAS číslo: 17407-56-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR