4-Methyl-2-pentenoic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6364-10G
OR6364-50G
APOSOR6364-10GEA
2700
CZK
APOSOR6364-10G
APOSOR6364-50G
4-Methyl-2-pentenoic acid 97%
4-Methyl-2-pentenoic acid
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Bod varu: 212 °C Hustota: 0,959 g/cm³ (25 °C) Bod vzplanutí: >100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043804 CAS číslo: 10321-71-8 EINECS: 233-706-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...