1-Benzylazetidine-3-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR6349-1G
OR6349-250MG
APOSOR6349-1GEA
7480
CZK
APOSOR6349-1G
APOSOR6349-250MG
1-Benzylazetidine-3-carboxylic acid 97%
1-Benzylazetidine-3-carboxylic acid
Vzorec:
C₁₁H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06752277 CAS číslo: 94985-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...