Tisk…

3-Methylenecyclobutanecarboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-methylidenecyclobutane-1-carboxylic acid

OR6346-1G OR6346-5G OR6346-250MG
APOSOR6346-1GEA 953 CZK
APOSOR6346-1G APOSOR6346-5G APOSOR6346-250MG
3-Methylenecyclobutanecarboxylic acid 97%
3-Methylenecyclobutanecarboxylic acid
Vzorec: C6H8O2
MW: 112.13 g/mol
Bod varu: 102…103 °C/8.3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00101901
CAS číslo: 15760-36-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR