5-Oxo-1-pyridin-4-ylpyrrolidine-3-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR6334-1G
OR6334-250MG
APOSOR6334-1GEA
17590
CZK
APOSOR6334-1G
APOSOR6334-250MG
5-Oxo-1-pyridin-4-ylpyrrolidine-3-carboxylic acid 95%
5-Oxo-1-pyridin-4-ylpyrrolidine-3-carboxylic acid
Vzorec:
C₁₀H₁₀N₂O₃ MW: 206,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07364770 CAS číslo: 914637-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...