Tisk…

Ethyl-3-furoate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl furan-3-carboxylate, Ethyl 3-Furoate, Ethyl 3-Furancarboxylate, 3-Furancarboxylic acid ethyl ester, 3-Furoic acid ethyl ester

OR6328-25G
APOSOR6328-25GEA 4900 CZK
APOSOR6328-25G
Ethyl-3-furoate 98%
Ethyl-3-furoate
Vzorec: C₇H₈O₃
MW: 140,14 g/mol
Bod varu: 93…95 °C/35mmHg
Hustota: 1,038 g/cm³ (25 °C)
Bod vzplanutí: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005348
CAS číslo: 614-98-2
EINECS: 210-403-5
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR