4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)-piperazine-2-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-((Benzyloxy)carbonyl)-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid, Piperazine-2-carboxylic acid, N1-BOC N4-CBZ protected
OR6323-1G
OR6323-5G
OR6323-250MG
OR6323-500MG
OR6323-10G
OR6323-25G
APOSOR6323-1GEA
3740
CZK
APOSOR6323-1G
APOSOR6323-5G
APOSOR6323-250MG
APOSOR6323-500MG
APOSOR6323-10G
APOSOR6323-25G
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)-piperazine-2-carboxylic acid 97%
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid
Vzorec:
C₁₈H₂₄N₂O₆ Bod tání: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683055 CAS číslo: 149057-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...