Tisk…

Piperidine-4-carboxylic acid hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: Isonipecotic acid hydrochloride

OR6322-100G OR6322-25G OR6322-5G
APOSOR6322-100GEA 22780 CZK
APOSOR6322-100G APOSOR6322-25G APOSOR6322-5G
Piperidine-4-carboxylic acid hydrochloride 95%
Piperidine-4-carboxylic acid hydrochloride
Vzorec: C₆H₁₂ClNO₂
MW: 165,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044736
CAS číslo: 5984-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR