Tisk…

1-Pyridin-2-ylpiperidine-4-carboxylic acid 98+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(Pyridin-2-yl)piperidine-4-carboxylic acid

OR6319-1G OR6319-5G
APOSOR6319-1GEA 0 CZK
APOSOR6319-1G APOSOR6319-5G
1-Pyridin-2-ylpiperidine-4-carboxylic acid 98+%
1-Pyridin-2-ylpiperidine-4-carboxylic acid
Vzorec: C₁₁H₁₄N₂O₂
MW: 206,25 g/mol
Bod tání: 152…154 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD06796180
CAS číslo: 685827-70-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR