Tisk…

1-(4-Methoxybenzyl)piperidine-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR6315-1G OR6315-5G OR6315-10G
APOSOR6315-1GEA 5490 CZK
APOSOR6315-1G APOSOR6315-5G APOSOR6315-10G
1-(4-Methoxybenzyl)piperidine-4-carboxylic acid
1-(4-Methoxybenzyl)piperidine-4-carboxylic acid
Vzorec: C₁₄H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD03371460
CAS číslo: 193538-22-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR