Tisk…

2,4-Dihydroxypyrimidine-5-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR6297-10G OR6297-1G OR6297-25G
APOSOR6297-10GEA 3940 CZK
APOSOR6297-10G APOSOR6297-1G APOSOR6297-25G
2,4-Dihydroxypyrimidine-5-carboxylic acid
2,4-Dihydroxypyrimidine-5-carboxylic acid
Vzorec: C₅H₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00006023
CAS číslo: 23945-44-0
EINECS: 245-947-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR