Tisk…

2-Amino-4-methylpyrimidine-5-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

OR6293-1G OR6293-250MG OR6293-5G
APOSOR6293-1GEA 1490 CZK
APOSOR6293-1G APOSOR6293-250MG APOSOR6293-5G
2-Amino-4-methylpyrimidine-5-carboxylic acid 95%
2-Amino-4-methylpyrimidine-5-carboxylic acid
Vzorec: C₆H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD05237187
CAS číslo: 769-51-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR