4-Oxazolecarboxylic acid 98+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Oxazole-4-carboxylic acid
OR6277-1G
OR6277-250MG
OR6277-5G
APOSOR6277-1GEA
622
CZK
APOSOR6277-1G
APOSOR6277-250MG
APOSOR6277-5G
4-Oxazolecarboxylic acid 98+%
4-Oxazolecarboxylic acid
Vzorec:
C₄H₃NO₃ MW: 113,0715 g/mol Bod tání: 130…135 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD06797151 CAS číslo: 23012-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...