4-Methylisoxazole-5-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR6276-1G
OR6276-250MG
APOSOR6276-1GEA
12370
CZK
APOSOR6276-1G
APOSOR6276-250MG
4-Methylisoxazole-5-carboxylic acid 95%
4-Methylisoxazole-5-carboxylic acid
Vzorec:
C₅H₅NO₃ Bod tání: 159…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235209 CAS číslo: 261350-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...