Tisk…

4-Aminopyridine-2-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Aminopyridine-2-carboxylic Acid

OR6260-1G OR6260-250MG
APOSOR6260-1GEA 2240 CZK
APOSOR6260-1G APOSOR6260-250MG
4-Aminopyridine-2-carboxylic acid 97%
4-Aminopyridine-2-carboxylic acid
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
Bod tání: 319 °C
Storage Temperature: Ambient
MDL Number: MFCD02685671
CAS číslo: 100047-36-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR