4-(Furan-2-yl)pyridine-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR6253-1G
OR6253-250MG
OR6253-500MG
APOSOR6253-1GEA
13630
CZK
APOSOR6253-1G
APOSOR6253-250MG
APOSOR6253-500MG
4-(Furan-2-yl)pyridine-2-carboxylic acid
4-(Furan-2-yl)pyridine-2-carboxylic acid
Vzorec:
C₁₀H₇NO₃ MW: 189,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08056303 CAS číslo: 914637-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...