Tisk…

3-(1H-Pyrazol-4-yl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

OR6231-1G OR6231-250MG OR6231-5G OR6231-500MG OR6231-2.5G
APOSOR6231-1GEA 3190 CZK
APOSOR6231-1G APOSOR6231-250MG APOSOR6231-5G APOSOR6231-500MG APOSOR6231-2.5G
3-(1H-Pyrazol-4-yl)benzoic acid 95%
3-(1H-Pyrazol-4-yl)benzoic acid
Vzorec: C₁₀H₈N₂O₂
MW: 188,1800 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06739075
CAS číslo: 1002535-21-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR