Tisk…

Cyclopropylboronic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR6228-25G OR6228-1G OR6228-5G OR6228-100G
APOSOR6228-25GEA 1820 CZK
APOSOR6228-25G APOSOR6228-1G APOSOR6228-5G APOSOR6228-100G
Cyclopropylboronic acid 98%
Cyclopropylboronic acid
Vzorec: C₃H₇BO₂
Bod tání: 90…95 °C
Storage Temperature: Freezer
MDL Number: MFCD04038750
CAS číslo: 411235-57-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR