3-Aminophenylboronic acid monohydrate 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate
OR6223-1G
OR6223-25G
OR6223-5G
APOSOR6223-1GEA
417
CZK
APOSOR6223-1G
APOSOR6223-25G
APOSOR6223-5G
3-Aminophenylboronic acid monohydrate 99%
3-Aminophenylboronic acid monohydrate
Vzorec:
C₆H₈BNO₂·1H₂O MW: 154.96 Bod tání: 93…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00149554 CAS číslo: 206658-89-1 EINECS: 250-189-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...