Benzylboronic acid 90%
Dodavatel: Apollo Scientific
|
Varování
|
OR6221-1G
OR6221-250MG
APOSOR6221-1GEA
11180
CZK
APOSOR6221-1G
APOSOR6221-250MG
Benzylboronic acid 90%
Benzylboronic acid
Vzorec:
C₇H₉BO₂ Bod tání: 161 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01114673 CAS číslo: 4463-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...