Tisk…

Acetobromo-α-D-glucuronic acid methyl ester 95+%

Dodavatel: Apollo Scientific
Varování

OR6200T-1G OR6200T-5G OR6200T-25G
APOSOR6200T-1GEA 1450 CZK
APOSOR6200T-1G APOSOR6200T-5G APOSOR6200T-25G
Acetobromo-α-D-glucuronic acid methyl ester 95+%
Acetobromo-alpha-D-glucuronic acid methyl ester
Vzorec: C₁₃H₁₇BrO₉
Bod tání: 82…110 °C
Storage Temperature: Freezer
MDL Number: MFCD00061613
CAS číslo: 21085-72-3
EINECS: 244-203-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR