Tisk…

3-(Aminomethyl)benzoic acid hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Aminomethyl)benzoic acid, HCl

OR6195-5G OR6195-25G
APOSOR6195-5GEA 1580 CZK
APOSOR6195-5G APOSOR6195-25G
3-(Aminomethyl)benzoic acid hydrochloride 95%
3-(Aminomethyl)benzoic acid hydrochloride
Vzorec: C₈H₁₀ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03791117
CAS číslo: 876-03-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR