Tisk…

2-Bromobenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (2-Bromophenyl)methanamine

OR6194-1G OR6194-25G OR6194-5G
APOSOR6194-1GEA 417 CZK
APOSOR6194-1G APOSOR6194-25G APOSOR6194-5G
2-Bromobenzylamine 98%
2-Bromobenzylamine
Vzorec: C₇H₈BrN
MW: 186.05
Bod varu: 118…119 °C/9mmHg
Storage Temperature: Ambient
MDL Number: MFCD00025572
CAS číslo: 3959-05-5
UN: 2735
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR