1H-Pyrrole-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Pyrrole-3-carboxaldehyde
OR6181-250MG
OR6181-10G
OR6181-1G
APOSOR6181-250MGEA
622
CZK
APOSOR6181-250MG
APOSOR6181-10G
APOSOR6181-1G
1H-Pyrrole-3-carbaldehyde 95%
1H-Pyrrole-3-carbaldehyde
Vzorec:
C₅H₅NO MW: 95,10 g/mol Bod tání: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426733 CAS číslo: 7126-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...