Tisk…

tert-Butyl-3-aminoazetidine-1-carboxylate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Amino-1-Boc-azetidine, tert-Butyl 3-aminoazetidine-1-carboxylate, 1-BOC-3-aminoazetidine

OR6152-25G OR6152-5G
APOSOR6152-25GEA 2400 CZK
APOSOR6152-25G APOSOR6152-5G
tert-Butyl-3-aminoazetidine-1-carboxylate 97%
tert-Butyl-3-aminoazetidine-1-carboxylate
Vzorec: C₈H₁₆N₂O₂
MW: 172,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861753
CAS číslo: 193269-78-2
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR