Tisk…

(S)-(-)-3-Aminoquinuclidine dihydrochloride 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

OR6149-25G
APOSOR6149-25GEA 5430 CZK
APOSOR6149-25G
(S)-(-)-3-Aminoquinuclidine dihydrochloride 99%
(S)-(-)-3-Aminoquinuclidine dihydrochloride
Vzorec: C₇H₁₆Cl₂N₂
MW: 199,1213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191753
CAS číslo: 119904-90-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR