Tisk…

N-Methyl-L-alanine 97%

Dodavatel: Apollo Scientific
Varování

OR61451-100G OR61451-25G
APOSOR61451-100GEA 3040 CZK
APOSOR61451-100G APOSOR61451-25G
N-Methyl-L-alanine 97%
N-Methyl-L-alanine
Vzorec: C₄H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00037241
CAS číslo: 3913-67-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR