Tisk…

2-Bromo-5-chloro-1,3-dimethylbenzene

Dodavatel: Apollo Scientific
Varování

OR61436-25G OR61436-5G OR61436-100G
APOSOR61436-25GEA 1910 CZK
APOSOR61436-25G APOSOR61436-5G APOSOR61436-100G
2-Bromo-5-chloro-1,3-dimethylbenzene
2-Bromo-5-chloro-1,3-dimethylbenzene
Vzorec: C₈H₈BrCl
Storage Temperature: Ambient
MDL Number: MFCD07780649
CAS číslo: 103724-99-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR