(R)-(+)-3-Aminoquinuclidine dihydrochloride 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR6142-1G
OR6142-25G
OR6142-5G
APOSOR6142-1GEA
662
CZK
APOSOR6142-1G
APOSOR6142-25G
APOSOR6142-5G
(R)-(+)-3-Aminoquinuclidine dihydrochloride 97%
(R)-(+)-3-Aminoquinuclidine dihydrochloride
Vzorec:
C₇H₁₆Cl₂N₂ Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191752 CAS číslo: 123536-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...