Benzyl-3-oxopyrrolidine-1-carboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Benzyloxycarbonyl-3-pyrrolidone, 1-Carbobenzoxy-3-pyrrolidone, 1-Cbz-3-pyrrolidinone, 1-Cbz-3-pyrrolidone, Benzyl 3-oxopyrrolidine-1-carboxylate, 1-Benzyloxycarbonyl-3-pyrrolidinone, 1-Carbobenzoxy-3-pyrrolidinone
OR61344-10G
OR61344-25G
APOSOR61344-10GEA
1990
CZK
APOSOR61344-10G
APOSOR61344-25G
Benzyl-3-oxopyrrolidine-1-carboxylate 97%
Benzyl-3-oxopyrrolidine-1-carboxylate
Vzorec:
C₁₂H₁₃NO₃ MW: 219,2365 g/mol Hustota: 1,1897 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03001711 CAS číslo: 130312-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...