6-Chloropyridazine-4-carboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR61342-10G
OR61342-1G
OR61342-5G
APOSOR61342-10GEA
19050
CZK
APOSOR61342-10G
APOSOR61342-1G
APOSOR61342-5G
6-Chloropyridazine-4-carboxylic acid 96%
6-Chloropyridazine-4-carboxylic acid
Vzorec:
C₅H₃ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD15146792 CAS číslo: 1256794-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...