Tisk…

tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate 97%

Dodavatel: Apollo Scientific

OR61341-5G OR61341-1G OR61341-10G
APOSOR61341-5GEA 2690 CZK
APOSOR61341-5G APOSOR61341-1G APOSOR61341-10G
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate
Vzorec: C₁₀H₂₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04115304
CAS číslo: 301673-16-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR