1,2-Diamino-3-chlorobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Chlorobenzene-1,2-diamine
OR61333-1G
OR61333-5G
OR61333-25G
APOSOR61333-1GEA
622
CZK
APOSOR61333-1G
APOSOR61333-5G
APOSOR61333-25G
1,2-Diamino-3-chlorobenzene 97%
1,2-Diamino-3-chlorobenzene
Vzorec:
C₆H₇ClN₂ MW: 142,59 g/mol Bod tání: 27…29 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09881742 CAS číslo: 21745-41-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...