1-Ethyl-5-methyl-1H-pyrazol-4-ylamine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amino-1-ethyl-5-methyl-1H-pyrazole hydrochloride, 1-Ethyl-5-methyl-1H-pyrazol-4-amine hydrochloride
OR61323-1G
OR61323-500MG
OR61323-250MG
APOSOR61323-1GEA
7210
CZK
APOSOR61323-1G
APOSOR61323-500MG
APOSOR61323-250MG
1-Ethyl-5-methyl-1H-pyrazol-4-ylamine hydrochloride
1-Ethyl-5-methyl-1H-pyrazol-4-ylamine hydrochloride
Vzorec:
C₆H₁₂ClN₃ MW: 161,64 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04968765 CAS číslo: 1185300-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...