Tisk…

tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl bis(2-hydroxyethyl)carbamate, N-BOC-Diethanolamine

OR61309-100G
APOSOR61309-100GEA 2820 CZK
APOSOR61309-100G
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate
Vzorec: C₉H₁₉NO₄
MW: 205,26 g/mol
Hustota: 1,085 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01862953
CAS číslo: 103898-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR