Tisk…

L-Proline-tert-butyl ester 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-tert-Butyl pyrrolidine-2-carboxylate, H-Pro-OtBu

OR61281-10G OR61281-25G OR61281-5G
APOSOR61281-10GEA 1830 CZK
APOSOR61281-10G APOSOR61281-25G APOSOR61281-5G
L-Proline-tert-butyl ester 97%
L-Proline-tert-butyl ester
Vzorec: C₉H₁₇NO₂
MW: 171,2 g/mol
Bod varu: 50…51 °C/1mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00037879
CAS číslo: 2812-46-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR