N-Benzylguanidinium acetate
Dodavatel: Apollo Scientific
OR61212-1G
OR61212-5G
APOSOR61212-1GEA
3540
CZK
APOSOR61212-1G
APOSOR61212-5G
N-Benzylguanidinium acetate
N-Benzylguanidinium acetate
Vzorec:
C₁₀H₁₅N₃O₂ MW: 209.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09864972 CAS číslo: 2211-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...