Tisk…

1H-Pyrazole-4-carbonitrile 98%

Dodavatel: Apollo Scientific
Varování

OR61205-5G OR61205-10G OR61205-1G
APOSOR61205-5GEA 1740 CZK
APOSOR61205-5G APOSOR61205-10G APOSOR61205-1G
1H-Pyrazole-4-carbonitrile 98%
1H-Pyrazole-4-carbonitrile
Vzorec: C₄H₃N₃
MW: 93,0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040250
CAS číslo: 31108-57-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR