6-Chloro-1H-pyrazolo[3,4-b]pyridine 98%
Dodavatel: Apollo Scientific
OR61201-10G
OR61201-1G
OR61201-5G
APOSOR61201-10GEA
25400
CZK
APOSOR61201-10G
APOSOR61201-1G
APOSOR61201-5G
6-Chloro-1H-pyrazolo[3,4-b]pyridine 98%
6-Chloro-1H-pyrazolo[3,4-b]pyridine
Vzorec:
C₆H₄ClN₃ MW: 153,5700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832900 CAS číslo: 63725-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...