Tisk…

6-Aminonicotinonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile

OR6119-100G OR6119-25G OR6119-5G
APOSOR6119-100GEA 15870 CZK
APOSOR6119-100G APOSOR6119-25G APOSOR6119-5G
6-Aminonicotinonitrile 97%
6-Aminonicotinonitrile
Vzorec: C₆H₅N₃
MW: 119,13 g/mol
Bod varu: 240…250 °C/15mmHg
Bod tání: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00128902
CAS číslo: 4214-73-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR