EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99%
Dodavatel: Apollo Scientific
Synonyms:
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate
OR61179-100G
OR61179-500G
OR61179-25G
APOSOR61179-100GEA
1290
CZK
APOSOR61179-100G
APOSOR61179-500G
APOSOR61179-25G
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99%
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Vzorec:
C₁₄H₁₇NO₃ MW: 247,29 g/mol Bod varu: 125…128 °C/0,1mmHg Bod tání: 63…66 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006703 CAS číslo: 16357-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...