Tisk…

1-(3-Methoxyphenyl)cyclopropanecarboxylic acid

Dodavatel: Apollo Scientific

OR61138-1G OR61138-100MG OR61138-250MG OR61138-5G
APOSOR61138-1GEA 2480 CZK
APOSOR61138-1G APOSOR61138-100MG APOSOR61138-250MG APOSOR61138-5G
1-(3-Methoxyphenyl)cyclopropanecarboxylic acid
1-(3-Methoxyphenyl)cyclopropanecarboxylic acid
Vzorec: C₁₁H₁₂O₃
MW: 192,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297151
CAS číslo: 74205-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR