3,5-Dimethylbenzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
α-Bromomesitylene, 1-(Bromomethyl)-3,5-dimethylbenzene
OR61128-100G
OR61128-25G
APOSOR61128-100GEA
6090
CZK
APOSOR61128-100G
APOSOR61128-25G
3,5-Dimethylbenzyl bromide 97%
3,5-Dimethylbenzyl bromide
Vzorec:
C₉H₁₁Br MW: 199,09 g/mol Bod varu: 109…110 °C/14mmHg Bod tání: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013539 CAS číslo: 27129-86-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...