5-Chloro-3-methylpyridine-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR61110-25G
OR61110-5G
APOSOR61110-25GEA
11540
CZK
APOSOR61110-25G
APOSOR61110-5G
5-Chloro-3-methylpyridine-2-carboxylic acid
5-Chloro-3-methylpyridine-2-carboxylic acid
Vzorec:
C7H6ClNO2 MW: 171,5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375127 CAS číslo: 886365-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...