tert-Butyl-N-methylcarbamate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR61100-5G
OR61100-25G
OR61100-100G
APOSOR61100-5GEA
622
CZK
APOSOR61100-5G
APOSOR61100-25G
APOSOR61100-100G
tert-Butyl-N-methylcarbamate 95%
tert-Butyl-N-methylcarbamate
Vzorec:
C₆H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08899404 CAS číslo: 16066-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...