Tisk…

Methyl-6-amino-5-bromopicolinate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 6-amino-5-bromopyridine-2-carboxylate, Methyl 6-amino-5-bromopicolinate

OR61091-1G OR61091-250MG OR61091-5G
APOSOR61091-1GEA 3230 CZK
APOSOR61091-1G APOSOR61091-250MG APOSOR61091-5G
Methyl-6-amino-5-bromopicolinate 97%
Methyl-6-amino-5-bromopicolinate
Vzorec: C₇H₇BrN₂O₂
MW: 231,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062950
CAS číslo: 178876-82-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR