2-Bromo-6-iodotoluene 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-3-iodo-2-methylbenzene
OR61090-10G
OR61090-1G
OR61090-5G
APOSOR61090-10GEA
4630
CZK
APOSOR61090-10G
APOSOR61090-1G
APOSOR61090-5G
2-Bromo-6-iodotoluene 98%
2-Bromo-6-iodotoluene
Vzorec:
C₇H₆BrI MW: 296,9300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780672 CAS číslo: 97456-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...