4-(Chloromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR61079-100G
OR61079-25G
OR61079-500G
APOSOR61079-100GEA
1040
CZK
APOSOR61079-100G
APOSOR61079-25G
APOSOR61079-500G
4-(Chloromethyl)benzonitrile
4-(Chloromethyl)benzonitrile
Vzorec:
C₈H₆ClN MW: 151.60 g/mol Bod varu: 179 °C/50mmHg Bod tání: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019761 CAS číslo: 874-86-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...