5-Bromo-1-isoindolinone 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2,3-dihydro-1H-isoindol-1-one, 5-Bromoisoindolin-1-one
OR61062-5G
OR61062-100G
OR61062-25G
APOSOR61062-5GEA
1040
CZK
APOSOR61062-5G
APOSOR61062-100G
APOSOR61062-25G
5-Bromo-1-isoindolinone 98%
5-Bromo-1-isoindolinone
Vzorec:
C₈H₆BrNO MW: 212,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09701292 CAS číslo: 552330-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...